
bisBenzimide H 33258 trihydrochloride (Hoechst 33258) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23491-45-4
:
Formula:C₂₅H₂₄N₆O·3HCl
MDL Number:MFCD00012679
:533.88 g/mol
EINECS:245-690-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:yellow powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Water:≤20 %
- Titration Argentometric:97.5 to 102.5 % (On dry substance)
Specification Test Results
Frequently Bought Together