Order Entry
Spain
ContactUsLinkComponent
2,3-Dimethylhexane 95%
2,3-Dimethylhexane 95%
New Product
Certificates
2,3-Dimethylhexane 95%
New Product

Specification Test Results

  • Appearance:
    clear colorless to almost colorless liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥94 %
  • Refractive index:
    1.3990 to 1.4010 (20°C, 589 nm)

Specification Test Results

Frequently Bought Together