5-Amino-1-naphthalenesulphonic acid tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:84-89-9
:
Formula:C₁₀H₉NO₃S
MDL Number:MFCD00014315
:223.25 g/mol
Merck Index:14,6404
Melting Pt:>300 °C
EINECS:201-571-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together