Order Entry
Spain
ContactUsLinkComponent
4-Amino-1-naphthalenesulphonic acid, Sigma-Aldrich®
Corrosive
4-Amino-1-naphthalenesulphonic acid, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
4-Amino-1-naphthalenesulphonic acid, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.31 g/l (20 °C)
  • Melting Point:
    300 °C
  • Molar Mass:
    223.24 g/mol
  • Density:
    1.67 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together