Order Entry
Spain
ContactUsLinkComponent
tiamo™ 3.0 Software, Full USB
tiamo™ 3.0 Software, Full USB
  METR66056302
 :  METROHM
New Product
tiamo™ 3.0 Software, Full USB
  METR66056302
 :  METROHM
New Product
  • Description:
    tiamo™ 3.0 software
  • Pk:
    1
Computer program for controlling complex titration systems.
  • Graphical method editor with numerous templates
  • Layout manager for individual monitor interface
  • Professional database with reevaluation
  • Export to LIMS, NuGenesis, Cyberlab, and more
  • High-performance report generator
  • Parallel titration
In compliance with the directives according to FDA 21 CFR Part 11.
Dialog languages: German, English, French, Italian, Spanish, Portuguese, Polish, Russian, Slovakian, Czech, Japanese, Chinese and Traditional Chinese.