3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] fluorescent dye
Catalog # STMC100-0815
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:572.531 g/mol
CAS Number:53213-82-4
Formula:C₂₉H₃₇IN₂O₂
MDL Number:MFCD00011954