Amplite® Colorimetric Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) Assay Kit
Catalog # AATB11321
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Pk:100 Tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® colorimetric GAPDH activity assay kit is a sensitive and simple tool for monitoring GAPDH activity based on a coupled enzymatic reaction catalysed by GAPDH.
- Robust protocol
- Absorbance measured at 450 nm
- Six month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Delivery information: Shipped with blue ice.
Caution: Research use only.