TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Catalog # AATB13206
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide name:TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Specifications
About this item
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2] has much stronger fluoresce upon protease cleavage than the commonly used FRET peptide MCA-HQKLVFFA (K-DNP), a secretase FRET peptide substrate.
- Stable for 12 months
- Optimised for FRET applications
Quality tested in-house.
Caution: For research use only.