Supelco® Organic Reference Standard, 4-Hexylresorcinol
93513-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:136-77-6
- Description:4-Hexylresorcinol
- Storage conditions:RT
- Hazard class description:H302 - H319 - H411
- Shipping conditions:STD
- MDL No.:MFCD00002284
- M-Clarity quality level:MQ100
- Synonym(s):4-Hexyl-1,3-dihydroxybenzene
- Melting point:65 - 67 °C (lit.)
- Formula:CH₃(CH₂)₅C₆H₃-1,3-(OH)₂
- Molecular weight:194,27
- Flash point:198 °C
- Boiling point:333 - 335 °C (lit.)
: P264 - P270 - P273 - P280 - P301 + P312 - P305 + P351 + P338