Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Entrada de pedido
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor® 680 tyramide
iFluor® 680 tyramide [Superior Replacement for Opal 690]
iFluor® 680 tyramide [Superior Replacement for Opal 690]
iFluor® 680 tyramide [Superior Replacement for Opal 690]
Catalog # AATB45113
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
iFluor® 680 tyramide [Superior Replacement for Opal 690]
Catalog # AATB45113
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Env:
200
Seq:
0200
Specifications
About this item
Product Details
:
Product Details