Order Entry
Spain
Orders LinkContactUsLinkComponent
Biotin azide

Biotin azide

5 mg
Catalog # AATB3023
Supplier:   AAT Bioquest
Biotin azide
Catalog # AATB3023
Supplier:  AAT Bioquest
CAS Number:  
Biotin azide

5 mg

CAS Number:  
Price
156,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:326.42 g/mol
Specifications

Specifications

  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.