Anti-ZDHHC15 Rabbit Polyclonal Antibody
Catalog # ANTIA100068-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:DHHC-15
- Antigen symbol:ZDHHC15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q96MV8
- Antigen synonyms:UNQ1969/PRO4501|ZDH15_HUMAN|Zinc finger DHHC domain-containing protein 15|Palmitoyltransferase ZDHHC15
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ZDHHC15 (amino acids 288 - 337)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ZDHHC15 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: ZDHHC15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human