Order Entry
Spain
ContactUsLinkComponent
 
Anti-Tcl1 Mouse Monoclonal Antibody [clone: ABT-TCL1]
New Product
 
undefined
Anti-Tcl1 Mouse Monoclonal Antibody [clone: ABT-TCL1]
New Product

 

  • Catalog No:
  • ANTIA100014-50
  • ANTIA100014-100
  • Antigen symbol:
  • Tcl1
  • Tcl1
  • Antigen name:
  • Anti TCL1A
  • Anti TCL1A
  • Antigen synonyms:
  • P14 TCL1|Protein p14 TCL1|Lymphoma/leukemia, T-cell|P14 TCL1 protein|Oncogene TCL1|T cell lymphoma 1|T cell leukemia 1|Oncogene TCL-1|T cell lymphoma 1A
  • P14 TCL1|Protein p14 TCL1|Lymphoma/leukemia, T-cell|P14 TCL1 protein|Oncogene TCL1|T cell lymphoma 1|T cell leukemia 1|Oncogene TCL-1|T cell lymphoma 1A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • ABT-TCL1
  • ABT-TCL1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# P56279
  • UniprotID# P56279
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide derived from human TCL1
  • Synthetic peptide derived from human TCL1
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Storage buffer:
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • IHC
  • IHC

 

Frequently Bought Together