Order Entry
Spain
ContactUsLinkComponent
Anti-CD70 Mouse Monoclonal Antibody [clone: TNFS7/1026]
Anti-CD70 Mouse Monoclonal Antibody [clone: TNFS7/1026]
  ANTIA253949-100
New Product
Anti-CD70 Mouse Monoclonal Antibody [clone: TNFS7/1026]
  ANTIA253949-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD 27L
  • Antigen symbol:
    CD70
  • Clonality:
    Monoclonal
  • Clone:
    TNFS7/1026
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P32970
  • Antigen synonyms:
    CD27LG|CD70 antigen|CD27-L|CD27 LG|CD 70|CD27 ligand|CD27L|CD70|CD27 L
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    30 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human CD70 protein.
  • Tested applications:
    Functional studies, IHC-Fr
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [TNFS7/1026] antibody to CD70 for ELISA, Functional Studies, Flow Cytometry, IF and IHC-Fr with samples derived from human.

  • Validated Applications: ELISA, Functional Studies, Flow Cytometry, IF, IHC to Fr
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml

Type: Primary
Antigen: CD70
Clonality: Monoclonal
Clone: TNFS7/1026
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human