Anti-Flt3/CD135 Rabbit Polyclonal Antibody
Catalog # ANTIA94932-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 135
- Antigen symbol:Flt3/CD135
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P36888
- Antigen synonyms:FL cytokine receptor|Fetal liver kinase 2|Flt3|Flt 3|Flk2|FLT-3|Flk 2|CD135|CD135 antigen
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:112 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic phospho-peptide derived from human FLT3 around the phosphorylation site of Tyr842
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to FLT3 (phospho Tyr842) for WB and ELISA with samples derived from Human and Mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Flt3/CD135
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse