Anti-LXR alpha + LXR beta Goat Polyclonal Antibody
Catalog # ANTIA83404-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Liver X receptor alpha
- Antigen symbol:LXR alpha + LXR beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:NR1H3|LXRbeta|LXRalpha|Nuclear orphan receptor LXR alpha|LXR b|NR1H2|Liver X receptor beta|LXR a|LXR alpha + LXR beta
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:CRLQDKKLPPLLSEI
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human NR1H3; NR1H2 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to LXR alpha + LXR beta for ELISA, WB, IF and flow cytometry with samples derived from human.
- Validated applications: ELISA, WB, IF, flow cytometry
Type: Primary
Antigen: LXR alpha + LXR beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human