Order Entry
Spain
ContactUsLinkComponent
 
Anti-eNOS Rabbit Polyclonal Antibody
  ANTIA10995-100
New Product
undefined
Anti-eNOS Rabbit Polyclonal Antibody
  ANTIA10995-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Endothelial nitric oxide synthase 3
  • Antigen symbol:
    eNOS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P29474
  • Antigen synonyms:
    Endothelial nitric oxidase synthase|Constitutive NOS|ecNOS|cNOS|eNOS|Endothelial nitric oxide synthase|EC NOS|EC-NOS|Endothelial NOS
  • Modification:
    phospho Ser1177
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    133 kDa
  • Sequence:
    TQSFS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic phosphorylated peptide around S1177 of human eNOS (NP_000594.2)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to eNOS (phospho Ser1177) for WB and IHC with samples derived from human and mouse.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: eNOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse