Anti-Ki67 Mouse Monoclonal Antibody [clone: MKI67/2462]
Catalog # ANTIA249345-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:Ki67
- Clonality:Monoclonal
- Clone:MKI67/2462
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P46013
- Antigen synonyms:MIB 1|MIB|Antigen KI-67|MKI67|KIA|Marker of proliferation Ki-67|KI67_HUMAN|Antigen KI67|PPP1R105
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:345/395 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 2293-2478, of human Ki67 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MKI67/2462] antibody to Ki67 for flow cytometry, IF, WB and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Ki67
Clonality: Monoclonal
Clone: MKI67/2462
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human