Order Entry
Spain
ContactUsLinkComponent
 
Mouse Ovalbumin Specific IgE ELISA Kit
  ANTIA74238-96
New Product
 :  
undefined
Mouse Ovalbumin Specific IgE ELISA Kit
  ANTIA74238-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Indirect
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Ovalbumin Specific IgE
  • Description:
    Mouse ovalbumin specific IgE ELISA kit
  • Environmentally Preferable:
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,563 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,938 ng/ml
  • Regulatory status:
    RUO

 

 

Mouse Ovalbumin Specific IgE ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Ovalbumin Specific IgE in serum, plasma, tissue homogenates, and other biological fluids.