Order Entry
Spain
ContactUsLinkComponent
 
Human GDF15 ELISA Kit
  ANTIA313794-96
New Product
 :  
undefined
Human GDF15 ELISA Kit
  ANTIA313794-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    GDF15
  • Description:
    Human GDF15 ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    100 - 1600 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    5,57 ng/L
  • Regulatory status:
    RUO

 

 

Human GDF15 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GDF15 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 100 to 1600 ng/L
  • Sensitivity: 5,57 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human GDF15 ELISA kit (A313794) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human GDF15 in serum, plasma or other biological fluids. An antibody specific for GDF15 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GDF15 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-GDF15 antibody, which also binds the GDF15 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-GDF15 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-GDF15 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of GDF15 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of GDF15 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.