- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:Cripto1/CRIPTO
- Description:Human Cripto1/CRIPTO ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:300 - 4800 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:13,96 ng/L
- Regulatory status:RUO
Human Cripto1/CRIPTO ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cripto1/CRIPTO in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 300 to 4800 ng/L
- Sensitivity: 13,96 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Cripto1/CRIPTO ELISA kit (A313415) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Cripto1/CRIPTO in serum, plasma or other biological fluids. An antibody specific for Cripto1/CRIPTO has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cripto1/CRIPTO present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Cripto1/CRIPTO antibody, which also binds the Cripto1/CRIPTO present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Cripto1/CRIPTO antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Cripto1/CRIPTO antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Cripto1/CRIPTO captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Cripto1/CRIPTO in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.