Order Entry
Spain
ContactUsLinkComponent
Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8] (FITC)
Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8] (FITC)
Certificates
undefined
Anti-CD41 Mouse Monoclonal Antibody [clone: HIP8] (FITC)

Specifications

  • Catalog No:
  • STMC60114FI
  • STMC60114FI.1
  • Antigen name:
  • CD41
  • CD41
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • HIP8
  • HIP8
  • Reactivity:
  • Baboon, Capuchin monkey, Pig, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
  • Baboon, Capuchin monkey, Pig, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3674
  • 3674
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together