Order Entry
Spain
ContactUsLinkComponent
 
Anti-SSEA-1 (CD15) Mouse Monoclonal Antibody [clone: MC-480] (Alexa Fluor® 488)
 
undefined
Anti-SSEA-1 (CD15) Mouse Monoclonal Antibody [clone: MC-480] (Alexa Fluor® 488)

 

  • Catalog No:
  • STMC60060AD.1
  • STMC60060AD
  • Antigen name:
  • SSEA-1 (CD15)
  • SSEA-1 (CD15)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 488
  • Alexa Fluor® 488
  • Clone:
  • MC-480
  • MC-480
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 14345
  • 14345
  • Isotype:
  • IgM kappa
  • IgM kappa

 

Frequently Bought Together