Order Entry
Spain
ContactUsLinkComponent
 
Anti-CD27 Hamster Monoclonal Antibody [clone: LG.3A10] (PE)
 
undefined
Anti-CD27 Hamster Monoclonal Antibody [clone: LG.3A10] (PE)

 

  • Catalog No:
  • STMC60160PE
  • STMC60160PE.1
  • Antigen name:
  • CD27
  • CD27
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • LG.3A10
  • LG.3A10
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Hamster
  • Hamster
  • Gene ID:
  • 21940/939/500318
  • 21940/939/500318
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together