Order Entry
Spain
ContactUsLinkComponent
Anti-CD45R (B220) Rat Monoclonal Antibody [clone: RA3-6B2] (Pacific Blue™)
Anti-CD45R (B220) Rat Monoclonal Antibody [clone: RA3-6B2] (Pacific Blue™)
Certificates
undefined
Anti-CD45R (B220) Rat Monoclonal Antibody [clone: RA3-6B2] (Pacific Blue™)

Specifications

  • Catalog No:
  • STMC60019PB.1
  • STMC60019PB
  • Antigen name:
  • CD45R (B220)
  • CD45R (B220)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Pacific Blue™
  • Pacific Blue™
  • Clone:
  • RA3-6B2
  • RA3-6B2
  • Reactivity:
  • Human, Mouse, Cat
  • Human, Mouse, Cat
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264/5788
  • 19264/5788
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together