Nitrite VHR L, 500 ml
Catalog # LOVI471170
Supplier: Lovibond
New Product
CAS Number:
Specifications
- Chemical method:M271
- Concentration:500 ml
- Pk:500 mL
- Description:Nitrite VHR L
- UN No.:3264
- Instrument:MD 600, MD 600 photometer series, MD 610, MD 640
Specifications
About this item
Easily monitor nitrite levels in closed-loop cooling systems with this liquid reagent. Proper control of nitrite levels prevents pipework corrosion. This easy-to-use colourimetric method test can monitor high levels (up to 2,500 mg/L) of nitrite without the need to dilute samples.
Unlike similar colourimetric tests on the market, this test provides the full testing range industrial water treaters need and avoids the release of potentially harmful nitrogen oxides during testing.