Order Entry
Spain
ContactUsLinkComponent
 
Anti-CD48 (SLAMF2) Hamster Monoclonal Antibody [clone: HM48-1] (APC)
 
undefined
Anti-CD48 (SLAMF2) Hamster Monoclonal Antibody [clone: HM48-1] (APC)

 

  • Catalog No:
  • STMC60162AZ.1
  • STMC60162AZ
  • Antigen name:
  • CD48 (SLAMF2)
  • CD48 (SLAMF2)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • HM48-1
  • HM48-1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Hamster
  • Hamster
  • Gene ID:
  • 12506
  • 12506
  • Isotype:
  • IgG1 lambda
  • IgG1 lambda

 

Frequently Bought Together