Order Entry
Spain
ContactUsLinkComponent
 
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE)
 
undefined
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PE)

 

  • Catalog No:
  • STMC60068PE
  • STMC60068PE.1
  • Antigen name:
  • CD44
  • CD44
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • IM7
  • IM7
  • Reactivity:
  • Baboon, Cat, Pig, Horse, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Bovine, Dog, Mouse, Squirrel monkey
  • Baboon, Cat, Pig, Horse, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Bovine, Dog, Mouse, Squirrel monkey
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12505/960
  • 12505/960
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa

 

Frequently Bought Together