Order Entry
Spain
ContactUsLinkComponent
Anti-Igg3 Isotype Control Mouse Monoclonal Antibody [clone: MG3-35]
Anti-Igg3 Isotype Control Mouse Monoclonal Antibody [clone: MG3-35]
Certificates
undefined
Anti-Igg3 Isotype Control Mouse Monoclonal Antibody [clone: MG3-35]

Specifications

  • Catalog No:
  • STMC60073.1
  • STMC60073
  • Antigen name:
  • Igg3 Isotype Control
  • Igg3 Isotype Control
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • MG3-35
  • MG3-35
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG3 kappa
  • IgG3 kappa

Specifications

Frequently Bought Together