Order Entry
Spain
ContactUsLinkComponent
Human Recombinant Heregulin-beta 1
Human Recombinant Heregulin-beta 1
Certificates
undefined
Human Recombinant Heregulin-beta 1

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    Heregulin-beta 1
  • Purity:
    ≥95%

Specifications

Frequently Bought Together