Order Entry
Spain
ContactUsLinkComponent
Mouse Recombinant IL-6
Mouse Recombinant IL-6
Certificates
undefined
Mouse Recombinant IL-6

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IL-6
  • Purity:
    ≥95%

Specifications

Frequently Bought Together