Order Entry
Spain
ContactUsLinkComponent
Human Recombinant IFN-beta (HEK293-expressed)
Human Recombinant IFN-beta (HEK293-expressed)
Certificates
undefined
Human Recombinant IFN-beta (HEK293-expressed)

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    B cell interferon|Fibroblast interferon|IFNB1|Leukocyte interferon|Type I interferon
  • Protein/peptide name:
    IFN-beta
  • Purity:
    ≥95%

Specifications

Frequently Bought Together