Order Entry
Spain
ContactUsLinkComponent
Human Recombinant Omentin
Human Recombinant Omentin
Catalog # STMC100-1325
CAS Number:  
undefined
Human Recombinant Omentin
Catalog # STMC100-1325
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    1286
  • Protein synonyms:
    Intelectin-1|ITLN-1
  • Protein/peptide name:
    Omentin
  • Purity:
    ≥90%

Specifications

About this item

Use omentin for the maintenance of insulin sensitivity and metabolism, decreasing levels of this adipokine have been associated with obesity, insulin resistance, and diabetes (Tan et al.). Studies have suggested that omentin may have protective effects against atherosclerosis, arterial calcification, and myocardial injury through AMP-activated protein kinase (AMPK) and Akt-dependent mechanisms (Xu et al., Kataoka et al.). Through regulation of Sirt1-dependent p53 deacetylation, omentin can inhibit proliferation and induce apoptosis in hepatocellular carcinoma cells (Zhang and Zhou). Omentin is mainly produced by visceral adipose tissue, but is also expressed in the small intestine, colon, mesothelial cells, vascular cells, and plasma (Watanabe et al). Protein sequence analysis has shown that omentin comprises 313 amino acids, with a secretory signal sequence and fibrinogen-related domain (Yang et al.). For consistency and reproducibility across your applications, Omentin from STEMCELL comes lyophilised with ≥90% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤5,0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').