Order Entry
Spain
ContactUsLinkComponent
MesenCult™ ACF Freezing Medium
MesenCult™ ACF Freezing Medium
Catalog # STMC05490
CAS Number:  
undefined
MesenCult™ ACF Freezing Medium
Catalog # STMC05490
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Description:
    MesenCult™-ACF Freezing Medium, 50 ml
  • Format:
    Human; animal component-free, serum-free

Specifications

About this item

Animal component-free MSC cryopreservation medium.

  • Serum-free and animal component-free
  • Reproducibly high recovery rates
  • Optimised for MSCs previously cultured in MesenCult™-ACF Plus medium or MesenCult™ medium
  • Preserves human MSC multipotency and expansion capacities
  • Convenient, ready-to-use format

MesenCult™-ACF Freezing medium is a defined, serum-free, and animal component-free medium for the cryopreservation of mesenchymal stem cells (MSCs). This complete and ready-to-use medium is recommended for human MSCs previously cultured in MesenCult™-ACF Plus medium or MesenCult™ medium (MesenCult™ proliferation kit). Frozen human MSCs should be stored at –135 °C (liquid nitrogen) or colder.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').