Order Entry
Spain
ContactUsLinkComponent
Human Recombinant IL-2 (E. coli expressed)
Human Recombinant IL-2 (E. coli expressed)
Certificates
undefined
Human Recombinant IL-2 (E. coli expressed)

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    IL-2
  • Purity:
    ≥95%

Specifications

Frequently Bought Together