Order Entry
Spain
ContactUsLinkComponent
 
Spectra™ Multicolor Broad Range Protein Ladder
  PIER26628
 :  
Spectra™ Multicolor Broad Range Protein Ladder
  PIER26628
 :  

 

  • Pk:
    250 µl
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Storage conditions:
    Upon receipt store at −20 ˚C
  • Protein/peptide name:
    Multicolor Broad Range Protein Ladder
  • Shipping temperature:
    Product is shipped with an ice pack.

 

 

Thermo Scientific Spectra Multicolor Low Range Protein Ladder is a mixture of six proteins and synthetic peptides (1,7 to 40 kDa) for use as size standards for peptides and small proteins in gel electrophoresis and western blotting.

  • Monitoring of protein migration during SDS-PAGE
  • Verifying western transfer efficiency
  • Approximate sizing of proteins on SDS-polyacrylamide gels and western blots
  • Locating a protein of interest for excision from an unstained preparative gel

The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

 : For Research Use Only. Not for use in diagnostic procedures.