Order Entry
Spain
ContactUsLinkComponent
PancreaCult™ Organoid Growth Medium (Human)
PancreaCult™ Organoid Growth Medium (Human)
Catálogo # STMC100-0781
Número CAS:  
undefined
PancreaCult™ Organoid Growth Medium (Human)
Catálogo # STMC100-0781
Número CAS:  
Especificaciones
  • Pk:
    1 KIT
  • Description:
    PancreaCult™ Organoid growth medium (human)
Especificaciones
Acerca de este artículo
Culture medium kit for growth, and establishment of human pancreatic organoids.
Reliably culture pancreatic duct organoids to study the function of the ductal epithelium, model pancreatic diseases such as cancer or cystic fibrosis, and perform targeted drug screens or toxicity assays. With the PancreaCult™ (Human) culture medium system, you have a complete and robust workflow to establish, expand, and maintain pancreatic duct organoids. Pancreatic organoids can be established from fresh or frozen dissociated tissue in PancreaCult™ Organoid Initiation Medium (OIM), optimised to provide higher rates of organoid formation from tissue. Organoids can then be cultured in PancreaCult™ Organoid Growth Medium (OGM) for long-term maintenance, passaging, serum-free drug screening, or cryopreservation for future experiments. The optional removal of EGF from the medium supports the depletion of normal cells in KRAS-activated cancer organoid cultures. Organoids grown using the PancreaCult™ (Human) medium system can be adapted to various culture protocols, including two-dimensional Transwell® monolayer cultures, dilute Corning® Matrigel® suspension cultures, and high-throughput assay-compatible culture formats. Learn how to set up human pancreatic organoids for drug testing in high-throughput formats using the latest protocol.
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').