Order Entry
Spain
ContactUsLinkComponent
RSeT™ Feeder-Free Medium
RSeT™ Feeder-Free Medium
Catalog # STMC05975
CAS Number:  
undefined
RSeT™ Feeder-Free Medium
Catalog # STMC05975
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    RSeT™ feeder-free medium
Specifications
About this item
Serum-free medium for naïve-like human pluripotent stem cells.
Revert primed human embryonic (ES) and induced pluripotent stem (iPS) cells to a naïve-like state and maintain cells in a naïve-like state under hypoxic conditions without bFGF or feeder cells. Developed under license from the Weizmann institute of science, this serum-free cell culture medium produces robust cultures with features of a naïve-like state, such as tightly packed, domed colonies with refractive edges. Human ES/iPS cells cultured in RSeT™ feeder-free ,medium show increased expression of key transcripts associated with the naïve-like state, such as KLF17, KLF2, KLF4, and TFCP2L1, and can either be differentiated or converted back to a primed state by culture in mTeSR™1 and then differentiated. Compatible products that can be used for differentiation include STEMdiff™ definitive endoderm kit, STEMdiff™ SMADi neural induction kit, and STEMdiff™ mesoderm induction medium.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').