Order Entry
Spain
Orders LinkContactUsLinkComponent
STEMdiff™ Neural Progenitor Medium
STEMdiff™ Neural Progenitor Medium
STEMdiff™ neural progenitor medium, 1 KIT
Catalog # STMC05833
Marketsource
STEMdiff™ Neural Progenitor Medium
Catalog # STMC05833
Marketsource
CAS Number:  
STEMdiff™ Neural Progenitor Medium
STEMdiff™ Neural Progenitor Medium
STEMdiff™ neural progenitor medium, 1 KIT
CAS Number:  
Price
516,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Medium for maintenance and expansion of neural progenitor cells derived from human ES and iPS cells.
STEMdiff™ neural progenitor medium is a defined and serum-free medium for the expansion of neural progenitor cells (NPCs) derived from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells using STEMdiff™ neural induction medium. NPCs cultured in this medium can be expanded 3 to 5 fold per passage, and cultured for at least 10 passages, with minimal spontaneous neuronal differentiation.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ neural progenitor medium