ClonaCell™ CHO ACF Medium
Catalog # STMC03816
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:90 mL
- Description:ClonaCell™-CHO ACF medium
Specifications
About this item
Serum-free, animal component-free semi-solid medium with supplements for selection and cloning of transfected CHO cells.
- Individual cells are suspended in viscous medium and form physically separated, discrete colonies that are easily isolated
- Monoclonal cell lines are isolated in less time using fewer resources compared with selection and cloning by limiting dilution
- Diverse clones with a wide range of growth rates and productivities form discrete colonies in the viscous medium
- Rare and high-producing clones can be individually isolated more easily using simultaneous selection and cloning in semi-solid medium compared with selection in bulk liquid cultures
ClonaCell™-CHO ACF is a methylcellulose-based semi-solid medium with supplements for the selection and cloning of suspension-adapted CHO cells in serum-free conditions. This medium is animal component-free and contains only recombinant proteins and synthetic components. Single-cell cloning efficiency of CHO cells is higher in this medium than in protein-free media, especially at low plating densities. It does not contain L-glutamine, selection agents, or phenol red and is compatible with dihydrofolate reductase (DHFR) and glutamine synthase (GS) selection systems.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').