Order Entry
Spain
ContactUsLinkComponent
Anti-CREB1 Rabbit Polyclonal Antibody
Anti-CREB1 Rabbit Polyclonal Antibody
Catalog # PRSI56-002
Supplier:  ProSci Inc.
New Product
undefined
Anti-CREB1 Rabbit Polyclonal Antibody
Catalog # PRSI56-002
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclic AMP-responsive element-binding protein 1
  • Antigen symbol:
    CREB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Epitope:
    Amino Acid Range = 105-132
  • Form:
    Liquid
  • Gene ID:
    P16220
  • Antigen synonyms:
    CREB1
  • Modification:
    None
  • Storage buffer:
    Supplied in PBS with 0,09% (W/V) sodium azide
  • Molecular weight:
    37 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This CREB1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 105-132 amino acids from the Central region of human CREB1.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Size:
    400 µl
  • Pk:
    400 µl

Specifications

About this item

This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in two transcript variants encoding different isoforms.

For FACS starting dilution is: 1:25 For WB starting dilution is: 1:1000. For IF starting dilution is: 1:10~50.

Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse, Rat