Anti-AMN1 Rabbit Polyclonal Antibody
400 µl
Catalog # PRSI55-498
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
AMN1 belongs to the AMN1 family. The exact function of AMN1 remains unknown.
For FACS starting dilution is: 1:10~50. For WB starting dilution is: 1:1000.
Type: Primary
Antigen: AMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Specifications
- Size:400 µl
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- Modification:None
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Antibody type:Primary
- Epitope:Amino Acid Range = 13-42
- Form:Liquid
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Host:Rabbit
- Concentration:0,31 mg/ml
- Molecular weight:28 kDa
- Shipping temperature:4 °C
- Antigen name:Protein AMN1 homolog
- Cat. No.:PRSI55-498
- Reactivity:Human
- Gene ID:Q8IY45
- Antigen synonyms:AMN1
- Immunogen:This AMN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 13-42 amino acids from the N-terminal region of human AMN1.
- Conjugation:Unconjugated
- Antigen symbol:AMN1
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide



