Specifications
- Antibody type:Primary
- Antigen name:alkaline phosphatase, liver/bone/kidney
- Antigen symbol:ALPL
- Clonality:Monoclonal
- Clone:ALPL/597
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P05186
- Antigen synonyms:ALPL
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant human ALPL protein was used as the immunogen for the Alkaline Phosphatase antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The first three are located together on chromosome 2, while the tissue non-specific form is located on chromosome 1. The product of this gene is a membrane bound glycosylated enzyme that is not expressed in any particular tissue and is, therefore, referred to as the tissue-nonspecific form of the enzyme. The exact physiological function of the alkaline phosphatases is not known. A proposed function of this form of the enzyme is matrix mineralization; however, mice that lack a functional form of this enzyme show normal skeletal development. This enzyme has been linked directly to hypo-phosphatasia, a disorder that is characterised by hypercalcemia and includes skeletal defects. The character of this disorder can vary, however, depending on the specific mutation since this determines age of onset and severity of symptoms. Alternatively spliced transcript variants, which encode the same protein, have been identified for this gene.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 1 - 2 µg/ml for 30 min at RT Prediluted format: incubate for 30 min at RT (1) Optimal dilution of the Alkaline Phosphatase antibody should be determined by the researcher.
1. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone: ALPL/597
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Bovine