Specifications
- Antibody type:Primary
- Antigen name:luteinizing hormone/choriogonadotropin receptor
- Antigen symbol:LHCGR
- Clonality:Monoclonal
- Clone:LHCGR/1416
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P22888
- Antigen synonyms:LHCGR
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:A partial recombinant protein from the N-terminal region of human Luteinizing hormone / Choriogonadotropin receptor was used as the immunogen for the hCG Receptor antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recognises a protein of 78-85 kDa, which is identified as the receptor for Luteinizing hormone/Choriogonadotropin. Luteinizing hormone plays a role in spermatogenesis and ovulation by stimulating the testes and ovaries to produce steroids. Choriogonadotropin production in the placenta maintains estrogen and progesterone levels during the first trimester of pregnancy. Ovaries and testes abundantly express luteinizing hormone/choriogonadotropin receptor (LHCGR) as a seven transmembrane, G protein-coupled receptor glycoprotein. LHCGR influences the protective effect of pregnancy and Gonadotropin against breast cancer. The expression of LHCGR on breast carcinoma correlates in part to the degree of tumor differentiation. LHCGR -positive breast tumors occur more frequently in tumors with greater cell differentiation in premenopausal women.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml IF: 1 - 2 µg/ml WB: 0,5 - 1 µg/ml Optimal dilution of the hCG Receptor antibody should be determined by the researcher.
Type: Primary
Antigen: LHCGR
Clonality: Monoclonal
Clone: LHCGR/1416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human