Anti-CLASRP Rabbit Polyclonal Antibody
PRSI61-896
New Product
- Antibody type:Primary
- Antigen name:CLK4-associating serine/arginine rich protein
- Antigen symbol:CLASRP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Western blot:Yes
- Epitope:254-283 aa
- Form:Liquid
- Gene ID:11129
- Modification:None
- Molecular weight:77 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SFRS16 antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 254-283 amino acids from the central region of human SFRS16.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: CLASRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 254-283
Host: Rabbit
Isotype: Ig
Reactivity: Human