Anti-CYB5R3 Rabbit Polyclonal Antibody
Catalog # PRSI23-012
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome b5 reductase 3
- Antigen symbol:CYB5R3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:1727
- Antigen synonyms:NADH-cytochrome b5 reductase 3|B5R|OTTHUMP00000198574|NADH-cytochrome b5 reductase 3 soluble form|OTTHUMP00000198665|DIA1|NADH-cytochrome b5 reductase 3 membrane-bound form|cytochrome b5 reductase 3|CYB5R3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 125-334 of human CYB5R3 (NP_001165131.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CYB5R3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat