Anti-TNFRSF4 Rabbit Polyclonal Antibody
PRSI22-987
New Product
- Antibody type:Primary
- Antigen name:TNF receptor superfamily member 4
- Antigen symbol:TNFRSF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:7293
- Antigen synonyms:OX40 cell surface antigen|OX40 antigen|IMD16|TAX transcriptionally-activated glycoprotein 1 receptor|ACT35|lymphoid activation antigene ACT35|ACT35 antigen|CD134 antigen|OX4TXGP1L|tax-transcriptionally activated glycoprotein 1 receptor|OX40L receptor|ATC35 antigen|OX40 homologue|CD134|tumor necrosis factor receptor superfamily member 4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 29-214 of human TNFRSF4 (NP_003318.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: TNFRSF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat