Anti-SUPT20H Rabbit Polyclonal Antibody
PRSI22-898
New Product
- Antibody type:Primary
- Antigen name:SPT20 homolog, SAGA complex component
- Antigen symbol:SUPT20H
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:55578
- Antigen synonyms:tumor rejection antigen|family with sequence similarity 48|FAM48A|protein FAM48A|C13|SPT2transcription factor SPT20 homolog|P38IP|suppressor of Ty 20 homolog|C13orf19|FP757|member A|transcription factor (p38 interacting protein)
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 580-779 of human SUPT20H (NP_001014308.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SUPT20H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat