Anti-MLKL Rabbit Polyclonal Antibody
PRSI19-745
New Product
- Antibody type:Primary
- Antigen name:Mixed lineage kinase domain-like protein
- Antigen symbol:MLKL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:74568
- Antigen synonyms:ECO:0000312|MGI:MGI:1921818}|Mlkl {ECO:0000312|EMBL:AAH237551|Mixed lineage kinase domain-like protein
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of mouse MLKL.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MLKL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse