Anti-PSMF1 Rabbit Polyclonal Antibody
Catalog # PRSI19-728
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) inhibitor subunit 1 (PI31)
- Antigen symbol:PSMF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:9491
- Antigen synonyms:proteasome inhibitor hP131 subunit|PSMF1|macropain) inhibitor subunit 1 (PI31)|proteasome inhibitor subunit 1|PI31|proteasome (prosome|RP4-545L17.1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-271 of human PSMF1 (NP_006805.2).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PSMF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat